ENA369382764 Enamine

3-iodoimidazo[1,2-a]pyridine-8-carboxylic acid

  • MDL number MFCD22056480
  • Cas Number : 1384429-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99357

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99357
Availability details for
Quantity
  • Availability
  • Add to Cart