ENA369382782 Enamine

5-(trifluoromethyl)-1-benzofuran

  • MDL number MFCD18413278
  • Cas Number : 741290-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-102974

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102974
Availability details for
Quantity
  • Availability
  • Add to Cart