ENA369382784 Enamine

1-(3,4-dimethylphenyl)-2-phenylethan-1-amine

  • MDL number MFCD11202643
  • Cas Number : 42825-57-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102977

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102977
Availability details for
Quantity
  • Availability
  • Add to Cart