ENA369382786 Enamine

N-(2-methoxyphenyl)piperidin-4-amine

  • MDL number MFCD12822084
  • Cas Number : 260360-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102982

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102982
Availability details for
Quantity
  • Availability
  • Add to Cart