ENA369382787 Enamine

1-{[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}-1H-pyrazole

  • MDL number MFCD20527118
  • Cas Number : 1486485-43-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102997

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102997
Availability details for
Quantity
  • Availability
  • Add to Cart