ENA369382799 Enamine

N-(2-chloroethyl)cyclopropanecarboxamide

  • MDL number MFCD11612173
  • Cas Number : 53487-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-103044

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103044
Availability details for
Quantity
  • Availability
  • Add to Cart