ENA369382801 Enamine

imidazo[1,2-a]pyridine-3-carbothioamide

  • MDL number MFCD20705308
  • Cas Number : 91982-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 214...216
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103047

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103047
Availability details for
Quantity
  • Availability
  • Add to Cart