ENA369382804 Enamine

1-aminobutan-2-one hydrochloride

  • MDL number MFCD13152262
  • Cas Number : 108661-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103052

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103052
Availability details for
Quantity
  • Availability
  • Add to Cart