ENA369382805 Enamine

3-methyl-4-(3-methylpiperidin-1-yl)benzonitrile

  • MDL number MFCD12766900
  • Cas Number : 1179754-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102464

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102464
Availability details for
Quantity
  • Availability
  • Add to Cart