ENA369382811 Enamine

1-[(1-aminobutan-2-yl)oxy]-3-fluorobenzene

  • MDL number MFCD06247650
  • Cas Number : 883539-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103068

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103068
Availability details for
Quantity
  • Availability
  • Add to Cart