ENA369382814 Enamine

2-(chloromethyl)-5-(4-chlorophenyl)-4-methyl-1,3-oxazole

  • MDL number MFCD11103565
  • Cas Number : 70886-70-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102535

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102535
Availability details for
Quantity
  • Availability
  • Add to Cart