ENA369382817 Enamine

5-amino-2-chloro-N-[(thiophen-2-yl)methyl]benzamide

  • MDL number MFCD11650815
  • Cas Number : 1099610-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-102537

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102537
Availability details for
Quantity
  • Availability
  • Add to Cart