ENA369382819 Enamine

2-(4-bromophenyl)acetamide

  • MDL number MFCD00126843
  • Cas Number : 74860-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192-194
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102539

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102539
Availability details for
Quantity
  • Availability
  • Add to Cart