ENA369382824 Enamine

(3-bromo-2-methoxyphenyl)methanamine hydrochloride

  • MDL number MFCD11045218
  • Cas Number : 1375997-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176...178
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102545

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102545
Availability details for
Quantity
  • Availability
  • Add to Cart