ENA369382831 Enamine

2-(2-methoxyethoxy)benzaldehyde

  • MDL number MFCD09036531
  • Cas Number : 92637-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-102549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102549
Availability details for
Quantity
  • Availability
  • Add to Cart