ENA369382834 Enamine

[3-(2-methoxyethoxy)phenyl]methanol

  • MDL number MFCD11149527
  • Cas Number : 911060-77-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-102550

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102550
Availability details for
Quantity
  • Availability
  • Add to Cart