ENA369382835 Enamine

2-(thiophen-3-yl)-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD14615196
  • Cas Number : 1206971-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107...109
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103157

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103157
Availability details for
Quantity
  • Availability
  • Add to Cart