ENA369382837 Enamine

4-[(6-methylpyrimidin-4-yl)oxy]aniline

  • MDL number MFCD09262552
  • Cas Number : 927982-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102551

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102551
Availability details for
Quantity
  • Availability
  • Add to Cart