ENA369382840 Enamine

N-(4-bromo-2-chlorophenyl)methanesulfonamide

  • MDL number MFCD05116782
  • Cas Number : 902092-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-103181

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103181
Availability details for
Quantity
  • Availability
  • Add to Cart