ENA369382859 Enamine

1-methyl-1H,2H,3H,4H-pyrido[3,4-b]pyrazin-3-one

  • MDL number MFCD21840753
  • Cas Number : 1384429-39-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 274...276
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103258

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103258
Availability details for
Quantity
  • Availability
  • Add to Cart