ENA369382860 Enamine

2-(methylsulfanyl)-4-phenyl-1H-imidazole

  • MDL number MFCD08050850
  • Cas Number : 25433-10-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103260

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103260
Availability details for
Quantity
  • Availability
  • Add to Cart