ENA369382866 Enamine

ethyl 2-[methyl(3-nitropyridin-4-yl)amino]acetate

  • MDL number MFCD15508987
  • Cas Number : 1384430-89-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103272

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103272
Availability details for
Quantity
  • Availability
  • Add to Cart