ENA369382872 Enamine

2-amino-2-(5-bromo-2-methoxyphenyl)acetic acid

  • MDL number MFCD02662392
  • Cas Number : 500696-01-5
  • Molecular Weight : 260.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-102574

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102574
Availability details for
Quantity
  • Availability
  • Add to Cart