ENA369382873 Enamine

3-fluoro-5-sulfamoylbenzoic acid

  • MDL number MFCD09044680
  • Cas Number : 926220-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103281

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103281
Availability details for
Quantity
  • Availability
  • Add to Cart