ENA369382877 Enamine

3-(2-methylphenyl)propan-1-ol

  • MDL number MFCD09028767
  • Cas Number : 14902-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103286

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103286
Availability details for
Quantity
  • Availability
  • Add to Cart