ENA369382881 Enamine

N-(3,3,3-trifluoropropyl)cyclopentanamine

  • MDL number MFCD12811534
  • Cas Number : 1183610-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103293

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103293
Availability details for
Quantity
  • Availability
  • Add to Cart