ENA369382883 Enamine

N-(3,3,3-trifluoropropyl)cyclohexanamine

  • MDL number MFCD12771232
  • Cas Number : 1178206-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103295

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103295
Availability details for
Quantity
  • Availability
  • Add to Cart