ENA369382887 Enamine

2,3-dihydro-1,3,4-oxadiazol-2-one

  • MDL number MFCD19673344
  • Cas Number : 51517-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103300

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103300
Availability details for
Quantity
  • Availability
  • Add to Cart