ENA369382888 Enamine

3-methyl-2-(1H-pyrrol-1-ylmethyl)pyridine

  • MDL number MFCD18327342
  • Cas Number : 1344291-01-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103301

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103301
Availability details for
Quantity
  • Availability
  • Add to Cart