ENA369382900 Enamine

2-(pyridin-4-yl)-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD11846891
  • Cas Number : 21346-36-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103439

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103439
Availability details for
Quantity
  • Availability
  • Add to Cart