ENA369382902 Enamine

3-benzyl-4-phenylbutanoic acid

  • MDL number MFCD12617162
  • Cas Number : 93651-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82...84
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103446

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103446
Availability details for
Quantity
  • Availability
  • Add to Cart