ENA369382905 Enamine

3-(1,3-thiazol-2-yloxy)phenol

  • MDL number MFCD12800360
  • Cas Number : 163298-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103455

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103455
Availability details for
Quantity
  • Availability
  • Add to Cart