ENA369382911 Enamine

methyl 2-[(6-chloropyrazin-2-yl)sulfanyl]acetate

  • MDL number MFCD12092398
  • Cas Number : 1156664-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 32...34
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103468

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103468
Availability details for
Quantity
  • Availability
  • Add to Cart