ENA369382917 Enamine

4-(4-aminophenoxy)-N,N-dimethylpyridine-2-carboxamide

  • MDL number MFCD12678173
  • Cas Number : 284462-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113...115
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103502

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103502
Availability details for
Quantity
  • Availability
  • Add to Cart