ENA369382925 Enamine

4-chloro-2-(methylamino)benzonitrile

  • MDL number MFCD17292320
  • Cas Number : 1369935-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-102466

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102466
Availability details for
Quantity
  • Availability
  • Add to Cart