ENA369382928 Enamine

5-chloro-2-(1,1-dioxo-1lambda6-thiomorpholin-4-yl)benzonitrile

  • MDL number MFCD17349887
  • Cas Number : 1271453-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103606

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103606
Availability details for
Quantity
  • Availability
  • Add to Cart