ENA369382929 Enamine

3-[5-(pyridin-3-yl)-1,3,4-oxadiazol-2-yl]propanoic acid

  • MDL number MFCD11107681
  • Cas Number : 933730-18-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197...199
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103607

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103607
Availability details for
Quantity
  • Availability
  • Add to Cart