ENA369382930 Enamine

1-oxo-2,3,4,5-tetrahydro-1H-2-benzazepine-8-sulfonyl chloride

  • MDL number MFCD09811209
  • Cas Number : 919091-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103608

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103608
Availability details for
Quantity
  • Availability
  • Add to Cart