ENA369382937 Enamine

2-[(7-chloroquinolin-4-yl)sulfanyl]ethan-1-ol

  • MDL number MFCD12169604
  • Cas Number : 856372-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-102476

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102476
Availability details for
Quantity
  • Availability
  • Add to Cart