ENA369382940 Enamine

4-(bromomethyl)-3-(difluoromethoxy)benzonitrile

  • MDL number MFCD18392196
  • Cas Number : 1261617-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102596

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102596
Availability details for
Quantity
  • Availability
  • Add to Cart