ENA369382943 Enamine

2-[4-(dimethylsulfamoyl)phenyl]propanoic acid

  • MDL number MFCD16248969
  • Cas Number : 1304886-37-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122...124
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102597

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102597
Availability details for
Quantity
  • Availability
  • Add to Cart