ENA369382944 Enamine

3-(3,5-dimethyl-1,2-oxazol-4-yl)propan-1-amine

  • MDL number MFCD06213632
  • Cas Number : 771572-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103650

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103650
Availability details for
Quantity
  • Availability
  • Add to Cart