ENA369382957 Enamine

2-(morpholin-3-yl)-1-(thiophen-2-yl)ethan-1-one hydrochloride

  • MDL number MFCD22196487
  • Cas Number : 1376271-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173...175
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102477

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102477
Availability details for
Quantity
  • Availability
  • Add to Cart