ENA369382959 Enamine

methyl 2-aminocyclohexane-1-carboxylate

  • MDL number MFCD08695014
  • Cas Number : 40015-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103744

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103744
Availability details for
Quantity
  • Availability
  • Add to Cart