ENA369382962 Enamine

2-(aminomethyl)-N,N,5-trimethylaniline

  • MDL number MFCD16704806
  • Cas Number : 1247238-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103747

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103747
Availability details for
Quantity
  • Availability
  • Add to Cart