ENA369382969 Enamine

3-amino-1,4-dimethyl-1,2-dihydropyridin-2-one

  • MDL number MFCD22196499
  • Cas Number : 1376294-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-102605

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102605
Availability details for
Quantity
  • Availability
  • Add to Cart