ENA369382970 Enamine

2-fluoro-N-{1,7,7-trimethylbicyclo[2.2.1]heptan-2-yl}pyridine-4-carboxamide

  • MDL number MFCD22196594
  • Cas Number : 1291647-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103773

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103773
Availability details for
Quantity
  • Availability
  • Add to Cart