ENA369382972 Enamine

tert-butyl(2-methanesulfonylethyl)amine

  • MDL number MFCD12767548
  • Cas Number : 1183755-41-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 39...41
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103782

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103782
Availability details for
Quantity
  • Availability
  • Add to Cart