ENA369382986 Enamine

N-(2-phenylethyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD19664340
  • Cas Number : 1342738-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103818

Safety Information

msds https://www.enaminestore.com/catalog/EN300-103818
Availability details for
Quantity
  • Availability
  • Add to Cart