ENA369382987 Enamine

{imidazo[1,2-a]pyridin-3-yl}methanol

  • MDL number MFCD09864457
  • Cas Number : 30489-43-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103848

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103848
Availability details for
Quantity
  • Availability
  • Add to Cart